Assay Details
| Gene Symbol: Acss3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108091.1 | NP_001101561.1 | 9 - 10 | 1436 | 72 | |
| XM_006241302.2 | XP_006241364.1 | 9 - 10 | 1434 | 72 | ||
| XM_006241303.2 | XP_006241365.1 | 9 - 10 | 2123 | 72 | ||
| XM_008765327.1 | XP_008763549.1 | 9 - 10 | 1174 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acss3
Genomic Map