Assay Details
| Gene Symbol: RGD1306474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108746.1 | NP_001102216.1 | 1 - 2 | 219 | 140 | |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1306474
Genomic Map