Assay Details
| Gene Symbol: Top1mt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002798.1 | NP_001002798.1 | 2 - 3 | 251 | 65 | |
| XM_006241784.2 | XP_006241846.1 | 2 - 3 | 451 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Top1mt
Genomic Map