Assay Details
| Gene Symbol: Zfp707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135903.1 | NP_001129375.1 | 1 | 230 | 66 | |
| XM_006241815.2 | XP_006241877.1 | 1 | 460 | 66 | ||
| XM_006241816.2 | XP_006241878.1 | 1 | 461 | 66 | ||
| GenBank mRNA | BC161959.1 | - | 1 | 230 | 9027463 | 66 |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp707
Genomic Map