Assay Details
Gene Symbol: Ly6e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017467.1 | NP_001017467.1 | 2 - 3 | 309 | 63 | |
XM_006241757.2 | XP_006241819.1 | 3 - 4 | 812 | 63 | ||
XM_006241759.2 | XP_006241821.1 | 2 - 3 | 449 | 63 | ||
XM_006241760.2 | XP_006241822.1 | 3 - 4 | 604 | 63 | ||
XM_006241762.2 | XP_006241824.1 | 2 - 3 | 672 | 63 | ||
XM_006241763.2 | XP_006241825.1 | 2 - 3 | 669 | 63 | ||
GenBank mRNA | BC079165.1 | - | 2 - 3 | 309 | 7129625 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
