Assay Details
| Gene Symbol: Tmprss6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130556.1 | NP_001124028.1 | 12 - 13 | 1672 | 58 | |
| XM_006241995.2 | XP_006242057.1 | 12 - 13 | 1689 | 58 | ||
| XM_006241996.2 | XP_006242058.1 | 12 - 13 | 1654 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P054676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133985 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P054676 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P054676 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P054676 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P20937 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10905295 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905304 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905294 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905302 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905298 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905305 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905303 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905297 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905296 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905301 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905291 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905290 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905300 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905293 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905292 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905306 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10905299 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10905289 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700027159.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027157.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027149.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027156.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027153.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027164.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027161.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027163.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027148.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027152.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027158.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027160.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027166.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027154.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027145.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027144.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027167.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027165.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027155.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027151.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027162.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700027169.rn.1 | TC0700003454.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10905289 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371265 | - | - |

Tmprss6
Genomic Map