Assay Details
| Gene Symbol: Grina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153308.4 | NP_695220.4 | 2 - 3 | 503 | 92 | |
| GenBank mRNA | BC062074.1 | - | 2 - 3 | 473 | 5599428 | 92 |
| CK475470.1 | - | 2 - 3 | 503 | 92 | ||
| S61973.1 | - | 2 - 3 | 453 | 92 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Grina
Genomic Map