Assay Details
Gene Symbol: Grina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153308.4 | NP_695220.4 | 5 - 6 | 935 | 59 | |
GenBank mRNA | BC062074.1 | - | 5 - 6 | 905 | 5599428 | 59 |
S61973.1 | - | 5 - 6 | 885 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
