Assay Details
| Gene Symbol: Nsmce2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024876.1 | NP_001020047.1 | 1 - 2 | 74 | 147 | |
| XM_006241671.2 | XP_006241733.1 | 1 - 2 | 114 | 147 | ||
| XM_008765506.1 | XP_008763728.1 | 1 - 2 | 114 | 147 | ||
| GenBank mRNA | BC097477.1 | - | 1 - 2 | 74 | 7458086 | 147 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC097477.1, NM_001024876.1, XM_006241671.2, XM_008765506.1 transcripts.
Set Membership: |
|

Nsmce2
Genomic Map