Assay Details
| Gene Symbol: Nsmce2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024876.1 | NP_001020047.1 | 4 - 5 | 499 | 94 | |
| XM_008765505.1 | XP_008763727.1 | 3 - 4 | 2659 | 94 | ||
| GenBank mRNA | BC097477.1 | - | 4 - 5 | 499 | 7458086 | 94 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nsmce2
Genomic Map