Assay Details
| Gene Symbol: A4galt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006242115.2 | XP_006242177.1 | 2 - 3 | 398 | 61 | |
| XM_006242117.2 | XP_006242179.1 | 2 - 3 | 274 | 61 | ||
| XM_006242120.1 | XP_006242182.1 | 2 - 3 | 161 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006242117.2, XM_006242120.1 transcripts.
Set Membership: |
|

A4galt
Genomic Map