Assay Details
Gene Symbol: Lrrc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025659.2 | NP_001020830.2 | 9 - 10 | 1233 | 64 | |
GenBank mRNA | BC097394.1 | - | 9 - 10 | 1234 | 7453303 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P015512 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P060069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722868 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577633 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144976 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P015512 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P060069 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P060069 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P015512 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P060069 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P015512 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47569 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78405 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 75737 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72243 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33267 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51497 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97232 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10904147 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10904144 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10904142 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10904143 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10904149 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10904141 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10904150 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10904145 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10904148 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10904146 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10904140 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10904139 | - | - |
Affymetrix Rat Clariom S Assay | PSR0700024869.rn.1 | TC0700003270.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700024870.rn.1 | TC0700003270.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10904139 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362612 | - | - |
