Assay Details
Gene Symbol: March9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100601.1 | NP_001094071.1 | 1 - 2 | 420 | 107 | |
GenBank mRNA | AB048842.1 | - | 1 - 2 | 420 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
