Assay Details
Gene Symbol: Syngr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019166.2 | NP_062039.1 | 1 - 2 | 152 | 87 | |
XM_006241971.2 | XP_006242033.1 | 1 - 2 | 157 | 87 | ||
GenBank mRNA | DN932005.1 | - | 1 - 2 | 114 | 87 | |
U39549.1 | - | 1 - 2 | 121 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
