Assay Details
| Gene Symbol: Syngr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019166.2 | NP_062039.1 | 2 - 3 | 386 | 68 | |
| XM_006241971.2 | XP_006242033.1 | 2 - 3 | 391 | 68 | ||
| GenBank mRNA | DN932005.1 | - | 2 - 3 | 348 | 68 | |
| DV724604.1 | - | 1 - 2 | 192 | 68 | ||
| U39549.1 | - | 2 - 3 | 355 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syngr1
Genomic Map