Assay Details
| Gene Symbol: Ppp1r16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130566.1 | NP_001124038.1 | 9 - 10 | 1842 | 64 | |
| XM_008765580.1 | XP_008763802.1 | 8 - 9 | 4316 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp1r16a
Genomic Map