Assay Details
| Gene Symbol: Uqcrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127553.2 | NP_001121025.1 | 1 - 2 | 40 | 61 | |
| GenBank mRNA | CB783276.1 | - | 1 - 2 | 40 | 61 | |
| FQ217669.1 | - | 1 - 2 | 31 | 61 | ||
| FQ217799.1 | - | 1 - 2 | 39 | 61 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001127553.2 transcripts.
This assay is in the 5'UTR region of the NM_001127553.2 transcripts.
Set Membership: |
|

Uqcrb
Genomic Map