Assay Details
| Gene Symbol: Apol2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241942.2 | XP_006242004.1 | 3 - 4 | 823 | 90 | |
| XM_006241943.2 | XP_006242005.1 | 4 - 5 | 705 | 90 | ||
| XM_006241944.2 | XP_006242006.1 | 3 - 4 | 540 | 90 | ||
| XM_006241945.1 | XP_006242007.1 | 3 - 4 | 276 | 90 | ||
| XM_006241946.2 | XP_006242008.1 | 3 - 4 | 520 | 90 | ||
| XM_006241947.2 | XP_006242009.1 | 4 - 5 | 596 | 90 | ||
| XM_006241948.2 | XP_006242010.1 | 4 - 5 | 568 | 90 | ||
| XM_008765644.1 | XP_008763866.1 | 3 - 4 | 296 | 90 | ||
| XM_008765645.1 | XP_008763867.1 | 4 - 5 | 548 | 90 | ||
| XM_235465.8 | XP_235465.6 | 4 - 5 | 726 | 90 | ||
| Gene Symbol: LOC100911804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750381.3 | XP_003750429.1 | 727 | 90 | ||
| XM_006241953.2 | XP_006242015.1 | 824 | 90 | |||
| XM_006241954.2 | XP_006242016.1 | 706 | 90 | |||
| XM_006241955.2 | XP_006242017.1 | 3 - 4 | 541 | 90 | ||
| XM_006241956.1 | XP_006242018.1 | 276 | 90 | |||
| XM_006241957.2 | XP_006242019.1 | 3 - 4 | 521 | 90 | ||
| XM_006241958.2 | XP_006242020.1 | 4 - 5 | 597 | 90 | ||
| XM_006241959.2 | XP_006242021.1 | 4 - 5 | 569 | 90 | ||
| XM_008765799.1 | XP_008764021.1 | 296 | 90 | |||
| XM_008765800.1 | XP_008764022.1 | 4 - 5 | 549 | 90 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Apol2
Genomic Map