Assay Details
| Gene Symbol: Npap60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012991.1 | NP_037123.1 | 6 - 7 | 1391 | 96 | |
| GenBank mRNA | BC097337.1 | - | 6 - 7 | 1109 | 7453939 | 96 |
| U41845.2 | - | 6 - 7 | 1391 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npap60
Genomic Map