Assay Details
Gene Symbol: Cacng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053351.1 | NP_445803.1 | 2 - 3 | 326 | 74 | |
GenBank mRNA | AF118818.1 | - | 2 - 3 | 326 | 74 | |
AF361339.1 | - | 2 - 3 | 293 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
neuromuscular junction development
transmission of nerve impulse
regulation of ion transmembrane transport
regulation of membrane potential
neurological system process
membrane depolarization
membrane hyperpolarization
calcium ion transmembrane transport
regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
voltage-gated calcium channel activity
calcium channel regulator activity
protein binding
ionotropic glutamate receptor binding
neuromuscular junction development
transmission of nerve impulse
regulation of ion transmembrane transport
regulation of membrane potential
neurological system process
membrane depolarization
membrane hyperpolarization
calcium ion transmembrane transport
regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
voltage-gated calcium channel activity
calcium channel regulator activity
protein binding
ionotropic glutamate receptor binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P146970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140574 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275886 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867983 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47249 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23531 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P146970 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P146970 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P146970 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P838347 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44125 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83652 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90912 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40007 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40928 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74467 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31969 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10905269 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10905268 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10905267 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10905266 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10905265 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10905264 | - | - |
Rat Genome 230 2.0 Array | 1368759_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368759_at | - | - |
Affymetrix HT Rat Focus Array | 1368759_at | - | - |
Affymetrix HT Rat Focus Array | 1368759_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700027083.rn.1 | TC0700003446.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700027084.rn.1 | TC0700003446.rn.1 | RTA TC |
Rat Expression Set 230 | 1368759_PM_at | - | - |
Rat Expression Set 230 | 1368759_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10905264 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22090380 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372288 | - | - |
RatRef-12 Expression BeadChip | ILMN_49899 | - | - |
