Assay Details
Gene Symbol: Wbp2nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001067725.4 | XP_001067725.2 | 1 - 2 | 88 | 102 | |
XM_008765810.1 | XP_008764032.1 | 1 - 2 | 88 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001067725.4, XM_008765810.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
