Assay Details
Gene Symbol: Wbp2nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001067725.4 | XP_001067725.2 | 2 - 3 | 198 | 150 | |
XM_008765810.1 | XP_008764032.1 | 2 - 3 | 198 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
