Assay Details
Gene Symbol: Ndufb9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127294.2 | NP_001120766.1 | 1 - 2 | 122 | 106 | |
GenBank mRNA | BC166927.1 | - | 1 - 2 | 122 | 6919700 | 106 |
BC168767.1 | - | 1 - 2 | 122 | 6891022 | 106 | |
FQ210477.1 | - | 1 - 2 | 129 | 106 | ||
FQ217145.1 | - | 1 - 2 | 128 | 106 | ||
FQ217579.1 | - | 1 - 2 | 141 | 106 | ||
FQ217728.1 | - | 1 - 2 | 122 | 106 | ||
FQ220460.1 | - | 1 - 2 | 141 | 106 | ||
FQ223611.1 | - | 1 - 2 | 141 | 106 | ||
FQ224000.1 | - | 1 - 2 | 187 | 106 | ||
FQ224618.1 | - | 1 - 2 | 158 | 106 | ||
FQ224680.1 | - | 1 - 2 | 129 | 106 | ||
FQ228493.1 | - | 1 - 2 | 129 | 106 | ||
FQ228673.1 | - | 1 - 2 | 141 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
