Assay Details
| Gene Symbol: Arhgap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004242.1 | NP_001004242.1 | 10 - 11 | 990 | 87 | |
| GenBank mRNA | BC079089.1 | - | 10 - 11 | 990 | 7123477 | 87 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P039342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72462 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435073 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121183 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P039342 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P039342 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P039342 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P492723 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66424 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75186 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 96212 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40994 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85867 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10898276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898277 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898275 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898271 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898280 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898269 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898278 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898272 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898279 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898270 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10898268 | - | - |
| Rat Genome 230 2.0 Array | 1376501_at | - | - |
| Rat Genome 230 2.0 Array | 1376501_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1376501_at | - | - |
| Affymetrix HT Rat Focus Array | 1376501_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0700028723.rn.1 | TC0700003624.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700028733.rn.1 | TC0700003624.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700028725.rn.1 | TC0700003624.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700028727.rn.1 | TC0700003624.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700028730.rn.1 | TC0700003624.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700028734.rn.1 | TC0700003624.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700028735.rn.1 | TC0700003624.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700028736.rn.1 | TC0700003624.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700028731.rn.1 | TC0700003624.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700028724.rn.1 | TC0700003624.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700028726.rn.1 | TC0700003624.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0700028732.rn.1 | TC0700003624.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376501_PM_at | - | - |
| Rat Expression Set 230 | 1376501_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10898268 | - | - |
| Rat Genome U34 Set | rc_AA945062_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367483 | - | - |
| RatRef-12 Expression BeadChip | ILMN_55601 | - | - |

Arhgap8
Genomic Map