Assay Details
Gene Symbol: Ccdc101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114502.1 | NP_001107974.1 | 8 - 9 | 792 | 109 | |
XM_006230231.2 | XP_006230293.1 | 8 - 9 | 898 | 109 | ||
XM_006230232.2 | XP_006230294.1 | 8 - 9 | 802 | 109 | ||
GenBank mRNA | FQ210727.1 | - | 8 - 9 | 767 | 109 | |
FQ232716.1 | - | 8 - 9 | 767 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P140776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511121 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32458 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P140776 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P140776 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P140776 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P994732 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74501 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20635 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10710880 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710877 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710876 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710881 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710883 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710885 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710878 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710879 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710882 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710884 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10710875 | - | - |
Rat Genome 230 2.0 Array | 1389676_at | - | - |
Rat Genome 230 2.0 Array | 1389676_PM_at | - | - |
Rat Genome 230 2.0 Array | 1395038_at | - | - |
Rat Genome 230 2.0 Array | 1395038_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1395038_at | - | - |
Affymetrix HT Rat Focus Array | 1389676_at | - | - |
Affymetrix HT Rat Focus Array | 1389676_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1395038_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100020088.rn.1 | TC0100002597.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100020085.rn.1 | TC0100002597.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100020086.rn.1 | TC0100002597.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100020087.rn.1 | TC0100002597.rn.1 | RTA TC |
Rat Expression Set 230 | 1389676_PM_at | - | - |
Rat Expression Set 230 | 1389676_at | - | - |
Rat Expression Set 230 | 1395038_PM_at | - | - |
Rat Expression Set 230 | 1395038_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710875 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21756403 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355554 | - | - |
