Assay Details
| Gene Symbol: Il4r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133380.2 | NP_596871.2 | 4 - 5 | 382 | 69 | |
| XM_006230215.2 | XP_006230277.1 | 5 - 6 | 533 | 69 | ||
| XM_006230216.2 | XP_006230278.1 | 3 - 4 | 323 | 69 | ||
| XM_008759862.1 | XP_008758084.1 | 4 - 5 | 5741 | 69 | ||
| XM_008759863.1 | XP_008758085.1 | 5 - 6 | 5688 | 69 | ||
| XM_008759864.1 | XP_008758086.1 | 5 - 6 | 5688 | 69 | ||
| GenBank mRNA | AB015746.1 | - | 4 - 5 | 389 | 69 | |
| AB015747.1 | - | 4 - 5 | 389 | 69 | ||
| X69903.1 | - | 220 | 69 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Il4r
Genomic Map