Assay Details
| Gene Symbol: Map6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017204.1 | NP_058900.1 | 3 - 4 | 2146 | 89 | |
| XM_006229755.2 | XP_006229817.1 | 2 - 3 | 2476 | 89 | ||
| GenBank mRNA | AJ002556.1 | - | 2 - 3 | 2165 | 89 | |
| BC078848.1 | - | 2 - 3 | 2024 | 89 | ||
| X93495.1 | - | 3 - 4 | 2146 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map6
Genomic Map