Assay Details
| Gene Symbol: Aldoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230209.2 | XP_006230271.1 | 3 - 4 | 390 | 77 | |
| XM_006230210.2 | XP_006230272.1 | 2 - 3 | 300 | 77 | ||
More Information
Set Membership: |
|

Aldoa
Genomic Map