Assay Details
Gene Symbol: Ddias | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126294.1 | NP_001119766.1 | 3 - 4 | 279 | 66 | |
GenBank mRNA | BC166470.1 | - | 3 - 4 | 279 | 9088340 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
