Assay Details
Gene Symbol: Olr231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001000206.1 | NP_001000206.1 | 1 | 561 | 76 | |
XM_008759709.1 | XP_008757931.1 | 2 | 1270 | 76 | ||
XM_008759710.1 | XP_008757932.1 | 3 | 1780 | 76 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
