Assay Details
Gene Symbol: Ints4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191629.1 | NP_001178558.1 | 16 - 17 | 2089 | 66 | |
GenBank mRNA | FQ218032.1 | - | 14 - 15 | 1742 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
