Assay Details
| Gene Symbol: Stard10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013069.1 | NP_001013087.1 | 3 - 4 | 565 | 91 | |
| XM_006229850.2 | XP_006229912.1 | 4 - 5 | 688 | 91 | ||
| XM_006229851.2 | XP_006229913.1 | 3 - 4 | 627 | 91 | ||
| GenBank mRNA | BC091411.1 | - | 3 - 4 | 565 | 7325640 | 91 |
More Information
Updated Mapping Information
Set Membership: |
|

Stard10
Genomic Map