Assay Details
| Gene Symbol: Zfp143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012169.1 | NP_001012169.1 | 15 - 16 | 1887 | 100 | |
| XM_006229994.1 | XP_006230056.1 | 15 - 16 | 1947 | 100 | ||
| GenBank mRNA | BC083578.1 | - | 15 - 16 | 1887 | 7127506 | 100 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P067460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672525 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52526 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P067460 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P067460 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P067460 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P243385 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85236 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21997 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85625 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45967 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10709841 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10709831 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10709842 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10709837 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10709829 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10709833 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10709838 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10709836 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10709832 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10709843 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10709835 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10709830 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10709839 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10709834 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10709840 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10709827 | - | - |
| Rat Genome 230 2.0 Array | 1378405_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1378405_at | - | - |
| Rat Genome 230 2.0 Array | 1379704_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1379704_at | - | - |
| Affymetrix HT Rat Focus Array | 1379704_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1378405_at | - | - |
| Affymetrix HT Rat Focus Array | 1379704_at | - | - |
| Affymetrix HT Rat Focus Array | 1378405_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100017824.rn.1 | TC0100002352.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100017825.rn.1 | TC0100002352.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100017823.rn.1 | TC0100002352.rn.1 | RTA TC |
| Rat Expression Set 230 | 1378405_PM_at | - | - |
| Rat Expression Set 230 | 1379704_at | - | - |
| Rat Expression Set 230 | 1378405_at | - | - |
| Rat Expression Set 230 | 1379704_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10709827 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21372383 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376551 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375799 | - | - |

Zfp143
Genomic Map