Assay Details
| Gene Symbol: Zfp143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012169.1 | NP_001012169.1 | 7 - 8 | 694 | 72 | |
| XM_006229994.1 | XP_006230056.1 | 7 - 8 | 754 | 72 | ||
| GenBank mRNA | BC083578.1 | - | 7 - 8 | 694 | 7127506 | 72 |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp143
Genomic Map