Assay Details
| Gene Symbol: Rras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013434.1 | NP_001013452.1 | 2 - 3 | 460 | 101 | |
| GenBank mRNA | BC091333.1 | - | 2 - 3 | 460 | 7314509 | 101 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rras2
Genomic Map