Assay Details
Gene Symbol: Olr59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173293.1 | NP_775415.1 | 1 - 2 | 579 | 88 | |
XM_008759669.1 | XP_008757891.1 | 2 - 3 | 88 | |||
XM_008759670.1 | XP_008757892.1 | 2 - 3 | 88 | |||
XM_008759671.1 | XP_008757893.1 | 2 - 3 | 88 | |||
GenBank mRNA | AF079864.1 | - | 1 - 2 | 579 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF079864.1, NM_173293.1, XM_008759669.1, XM_008759670.1, XM_008759671.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
