Assay Details
Gene Symbol: Tbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106786.1 | NP_001100256.1 | 15 - 16 | 1822 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
activation of innate immune response
protein phosphorylation
negative regulation of gene expression
positive regulation of interferon-alpha production
positive regulation of interferon-beta production
positive regulation of I-kappaB kinase/NF-kappaB signaling
dendritic cell proliferation
positive regulation of interferon-beta biosynthetic process
positive regulation of transcription from RNA polymerase II promoter
defense response to Gram-positive bacterium
nucleic acid binding
protein kinase activity
ATP binding
phosphoprotein binding
activation of innate immune response
protein phosphorylation
negative regulation of gene expression
positive regulation of interferon-alpha production
positive regulation of interferon-beta production
positive regulation of I-kappaB kinase/NF-kappaB signaling
dendritic cell proliferation
positive regulation of interferon-beta biosynthetic process
positive regulation of transcription from RNA polymerase II promoter
defense response to Gram-positive bacterium
nucleic acid binding
protein kinase activity
ATP binding
phosphoprotein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P424541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1679 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P424541 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P424541 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P424541 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17992 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P424541 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64043 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10902715 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902727 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902723 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902728 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902721 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902726 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902722 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902719 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902725 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902731 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902717 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902732 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902716 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902729 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902724 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902720 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902718 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902733 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902730 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902734 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10902714 | - | - |
Rat Genome 230 2.0 Array | 1376880_at | - | - |
Rat Genome 230 2.0 Array | 1376880_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376880_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376880_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700021880.rn.1 | TC0700002822.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700021879.rn.1 | TC0700002822.rn.1 | RTA TC |
Rat Expression Set 230 | 1376880_at | - | - |
Rat Expression Set 230 | 1376880_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10902714 | - | - |
Rat Genome U34 Set | rc_AI639447_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20727209 | - | - |
ABI Rat Genome Survey Microarray | 21042392 | - | - |
ABI Rat Genome Survey Microarray | 21502232 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367327 | - | - |
