Assay Details
| Gene Symbol: Mrpl42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106782.1 | NP_001100252.1 | 2 - 3 | 187 | 150 | |
| XM_008765315.1 | XP_008763537.1 | 2 - 3 | 193 | 150 | ||
| GenBank mRNA | BC168747.1 | - | 2 - 3 | 175 | 6920919 | 150 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrpl42
Genomic Map