Assay Details
Gene Symbol: Chst11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108079.1 | NP_001101549.1 | 1 - 2 | 482 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chondrocyte development
respiratory gaseous exchange
post-embryonic development
carbohydrate biosynthetic process
chondroitin sulfate metabolic process
chondroitin sulfate biosynthetic process
embryonic limb morphogenesis
negative regulation of transforming growth factor beta receptor signaling pathway
polysaccharide localization
post-anal tail morphogenesis
regulation of cell proliferation
embryonic digit morphogenesis
negative regulation of apoptotic process
developmental growth
embryonic viscerocranium morphogenesis
embryonic skeletal system morphogenesis
cartilage development
N-acetylgalactosamine 4-O-sulfotransferase activity
sulfotransferase activity
chondroitin 4-sulfotransferase activity
N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase activity
chondrocyte development
respiratory gaseous exchange
post-embryonic development
carbohydrate biosynthetic process
chondroitin sulfate metabolic process
chondroitin sulfate biosynthetic process
embryonic limb morphogenesis
negative regulation of transforming growth factor beta receptor signaling pathway
polysaccharide localization
post-anal tail morphogenesis
regulation of cell proliferation
embryonic digit morphogenesis
negative regulation of apoptotic process
developmental growth
embryonic viscerocranium morphogenesis
embryonic skeletal system morphogenesis
cartilage development
N-acetylgalactosamine 4-O-sulfotransferase activity
sulfotransferase activity
chondroitin 4-sulfotransferase activity
N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P128209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462369 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905010 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174597 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244306 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629015 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 666259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929633 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68012 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 83684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21040 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P128209 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P128209 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P128209 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P101919 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97813 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 52660 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92650 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 98070 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4055 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10901412 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901411 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901410 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10901409 | - | - |
Rat Genome 230 2.0 Array | 1381767_at | - | - |
Rat Genome 230 2.0 Array | 1381767_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700018925.rn.1 | TC0700002391.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700018927.rn.1 | TC0700002391.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700018924.rn.1 | TC0700002391.rn.1 | RTA TC |
Rat Expression Set 230 | 1381767_at | - | - |
Rat Expression Set 230 | 1381767_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10901409 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21417670 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368516 | - | - |
