Assay Details
Gene Symbol: Cdk17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108082.2 | NP_001101552.2 | 3 - 4 | 641 | 80 | |
XM_006241253.2 | XP_006241315.1 | 3 - 4 | 891 | 80 | ||
GenBank mRNA | AB005540.1 | - | 3 - 4 | 606 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P14690 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P146682 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P086550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680226 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138515 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14690 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P146682 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P086550 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P146682 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P14690 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P086550 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P14690 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P146682 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P086550 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P689357 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P14690 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700482576 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700517587 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3983 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700517587 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700482576 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10894872 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894869 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894871 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894873 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894868 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894864 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894865 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894867 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894861 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894858 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894866 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894860 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894863 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894870 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894859 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894862 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10894857 | - | - |
Rat Genome 230 2.0 Array | 1374134_at | - | - |
Rat Genome 230 2.0 Array | 1374134_PM_at | - | - |
Rat Genome 230 2.0 Array | 1394473_at | - | - |
Rat Genome 230 2.0 Array | 1382345_at | - | - |
Rat Genome 230 2.0 Array | 1394473_PM_at | - | - |
Rat Genome 230 2.0 Array | 1382345_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382345_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382345_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700004214.rn.1 | TC0700000505.rn.1 | RTA TC |
Rat Expression Set 230 | 1382345_at | - | - |
Rat Expression Set 230 | 1382345_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10894857 | - | - |
Rat Genome U34 Set | AB005540_g_at | - | - |
Rat Genome U34 Set | AB005540_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22353869 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373247 | - | - |
