Assay Details
| Gene Symbol: Caps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108990.1 | NP_001102460.1 | 6 - 7 | 616 | 76 | |
| XM_006241337.2 | XP_006241399.1 | 16 - 17 | 1964 | 76 | ||
| XM_008765359.1 | XP_008763581.1 | 15 - 16 | 1496 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Caps2
Genomic Map