Assay Details
Gene Symbol: Rap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134346.3 | NP_599173.2 | 3 - 4 | 186 | 84 | |
XM_008765377.1 | XP_008763599.1 | 4 - 5 | 746 | 84 | ||
GenBank mRNA | AY607847.1 | - | 3 - 4 | 186 | 84 | |
BC081731.1 | - | 3 - 4 | 331 | 7192310 | 84 | |
D88313.1 | - | 3 - 4 | 158 | 84 | ||
FQ229455.1 | - | 3 - 4 | 360 | 84 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
