Assay Details
| Gene Symbol: Helb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006241426.2 | XP_006241488.1 | 11 - 12 | 2990 | 63 | |
| XM_006241428.2 | XP_006241490.1 | 10 - 11 | 2743 | 63 | ||
More Information
Set Membership: |
|

Helb
Genomic Map