Assay Details
Gene Symbol: RGD1311703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230086.2 | XP_006230148.1 | 1 - 2 | 211 | 110 | |
GenBank mRNA | BC089924.1 | - | 1 - 2 | 160 | 7307309 | 110 |
FM053223.1 | - | 1 - 2 | 215 | 110 | ||
FQ225875.1 | - | 1 - 2 | 211 | 110 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC089924.1, XM_006230086.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
