Assay Details
Gene Symbol: RGD1311703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230086.2 | XP_006230148.1 | 2 - 3 | 301 | 122 | |
GenBank mRNA | FM053223.1 | - | 2 - 3 | 305 | 122 | |
FQ225875.1 | - | 2 - 3 | 301 | 122 | ||
FQ227306.1 | - | 2 - 3 | 301 | 122 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
