Assay Details
| Gene Symbol: Acsm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014162.4 | NP_001014184.3 | 2 - 3 | 452 | 69 | |
| XM_006230123.1 | XP_006230185.1 | 3 - 4 | 395 | 69 | ||
| XM_006230126.1 | XP_006230188.1 | 3 - 4 | 395 | 69 | ||
| GenBank mRNA | AY325244.1 | - | 688 | 69 | ||
| BC078915.1 | - | 2 - 3 | 451 | 7096091 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acsm5
Genomic Map