Assay Details
| Gene Symbol: Eif3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277302.1 | NP_001264231.1 | 2 - 3 | 472 | 132 | |
| GenBank mRNA | FQ212642.1 | - | 2 - 3 | 461 | 132 | |
| FQ213714.1 | - | 2 - 3 | 472 | 132 | ||
| FQ229379.1 | - | 2 - 3 | 146 | 132 | ||
| FQ230130.1 | - | 2 - 3 | 460 | 132 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif3f
Genomic Map