Assay Details
Gene Symbol: Taf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134735.1 | NP_001128207.1 | 2 - 3 | 407 | 76 | |
GenBank mRNA | BC168203.1 | - | 2 - 3 | 407 | 7115651 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
