Assay Details
Gene Symbol: Sult1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031834.1 | NP_114022.1 | 2 - 3 | 205 | 158 | |
GenBank mRNA | AF394783.1 | - | 1 - 2 | 128 | 158 | |
FQ209630.1 | - | 2 - 3 | 214 | 158 | ||
FQ210886.1 | - | 2 - 3 | 210 | 158 | ||
FQ214244.1 | - | 2 - 3 | 210 | 158 | ||
FQ218459.1 | - | 2 - 3 | 210 | 158 | ||
FQ218506.1 | - | 2 - 3 | 219 | 158 | ||
FQ218658.1 | - | 2 - 3 | 209 | 158 | ||
FQ219178.1 | - | 2 - 3 | 210 | 158 | ||
FQ219521.1 | - | 2 - 3 | 210 | 158 | ||
FQ219542.1 | - | 2 - 3 | 210 | 158 | ||
FQ219948.1 | - | 2 - 3 | 210 | 158 | ||
L19998.1 | - | 2 - 3 | 205 | 158 | ||
X52883.1 | - | 2 - 3 | 193 | 158 | ||
X68640.1 | - | 1 - 2 | 110 | 158 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
