Assay Details
| Gene Symbol: Sbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134970.1 | NP_001128442.1 | 30 - 31 | 4173 | 58 | |
| XM_008759797.1 | XP_008758019.1 | 32 - 33 | 4485 | 58 | ||
| XM_008759798.1 | XP_008758020.1 | 32 - 33 | 4482 | 58 | ||
| XM_008759799.1 | XP_008758021.1 | 31 - 32 | 4410 | 58 | ||
| XM_008759800.1 | XP_008758022.1 | 31 - 32 | 4389 | 58 | ||
| XM_008759801.1 | XP_008758023.1 | 31 - 32 | 4386 | 58 | ||
| XM_008759802.1 | XP_008758024.1 | 31 - 32 | 4347 | 58 | ||
| XM_008759803.1 | XP_008758025.1 | 31 - 32 | 4265 | 58 | ||
| XM_008759804.1 | XP_008758026.1 | 30 - 31 | 4314 | 58 | ||
| GenBank mRNA | BC168698.1 | - | 30 - 31 | 4173 | 7938988 | 58 |
More Information
Updated Mapping Information
Set Membership: |
|

Sbf2
Genomic Map